2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazin ≥97%

Lieferant: Thermo Scientific
Achtung

B25742.03
B25742.03EA 62.7 EUR
B25742.03
2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazin ≥97%
2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazin

Purity: 97% CAS No.: 43167-79-9 Molecular Formula: C19H20N6O Formula Weight: 348.41 Melting Point: 194-196° MDL No.: MFCD00272362

Formel: C₁₉H₂₀N₆O
Molecular Weight: 348,41 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272362
CAS-Nummer: 43167-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR