L-Arginin Hydrochlorid 98,0-102,0% (durch titrimetrische Analyse an Trockenmasse), BAKER ANALYZED®, Biochemiereagenz, J.T.Baker®

Lieferant: Avantor

Synonyme: L(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, L(+)-Arginin Hydrochlorid, H-Arg-OH.HCl, (S)-(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, (S)-(+)-Arginin Hydrochlorid, H-L-Arg-OH.HCl

B577-05EA 211.13 EUR
B577-05 B577-06
L-Arginin Hydrochlorid 98,0-102,0% (durch titrimetrische Analyse an Trockenmasse), BAKER ANALYZED®, Biochemiereagenz, J.T.Baker®
L-Arginin Hydrochlorid
Formel: C₆H₁₅ClN₄O₂
Schmelzpunkt: 218 °C
Dichte: 1,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064550
CAS-Nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₄N₄O₂HCl) (dried basis, by non-aqueous acid- base titration) 98.0 - 102.0 %
Specific Rotation,α²ºD(dried basis, c=80mg in 6N HCL)(+) 21.4 - 23.6 Degree
Homogeneity by TLC Passes Test
Ash (sulfated) ≤ 0.1 %
Loss on Drying at 105°C ≤ 0.5 %
Heavy Metals (as Pb) ≤ 0.002 %
Iron (Fe) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR