Mag-Bind® Fit24™ Blood and Tissue DNA Kit
Lieferant: OMEGA BIO-TEK
Pre-scripted solution for purification of high-quality DNA from blood, saliva, cultured cells, or fresh or frozen tissue.
- Ease-of-use, automation-ready cartridges pre-filled with reagents
- Convenience, no reagent preparation and buffer dispensing
- High-quality purification, high-quality DNA from blood, saliva, fresh or frozen animal cultured cells, and tissue
Unleash the potential of your DNA purification with the MB Fit24™ Blood and Tissue DNA Kit - a convenient solution for rapid and reliable genomic DNA isolation. Tailored for diverse samples, including blood, saliva, fresh or frozen animal cultured cells, and tissues, this kit seamlessly integrates with the MagBinder® Fit24 Nucleic Acid Purification System.
Experience increased efficiency with the MB Fit24™ Blood and Tissue DNA Kit. Designed for automation, this kit comes with pre-filled reagent cartridges specifically configured for the MagBinder® Fit24 platform. Enjoy faster and consistent results with a semi-automated extraction workflow capable of processing up to 24 samples in less than 50 minutes.
Elevate workflows by eliminating reagent preparation and buffer dispensing steps. The MB Fit24™ Blood and Tissue DNA Kit ensures ease of use and convenience. Samples are lysed offline, and the lysate seamlessly transfers to the designated reagent well containing the binding buffer.
Experience the power of Mag-Bind® Particles PF-HDQ, providing reversible nucleic acid binding properties. Paired with Omega Bio-tek’s buffers, this system offers a fast and convenient method for isolating DNA from diverse sample types.
The purification procedure delivers high-quality DNA, ready for direct use in various downstream applications such as PCR amplification, next-generation sequencing, and enzymatic reactions.