Coomassie® Brillantblau R 250 Proteinfärbemittel, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83

M-Clarity™ Qualitätsstufe = MQ200

B6529-1L
B6529-1LEA 290 EUR
B6529-1L
Coomassie® Brillantblau R 250 Proteinfärbemittel, Sigma-Aldrich®
Coomassie® Brillantblau R 250

Brilliant Blue R staining solution is especially designed for use in staining protein in agarose gels following immunoelectrophoresis and Ouchterlony gels. The stain contains ethanol and acetic acid so gels do not require fixing prior to staining. The immunoelectrophoresis or Ouchterlony gel is first washed with water and saline to remove non-precipitated proteins and then dried. The gel is then immersed in staining solution for 30 min and destained with 10% acetic acid. For detection of protein bands following electrophoresis.

Formel: C₄₅H₄₄N₃NaO₇S₂
Molecular Weight: 825,98 g/mol
Flammpunkt: 27,2 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00041762
CAS-Nummer: 6104-59-2
UN: 1170
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=099b3b51de7b923ec416fec18e51652818a42f54&vid=e0beef0dea0843361ad24fbb11c007a88e813af7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Suitability for immunoelectrophoresis protein staining Pass
Appearance (Colour) Dark Blue
Appearance (Form) Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR