BCECF AM ester 1 mg/mL in DMSO fluoreszierender pH-Indikator, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
B8806-1MG
B8806-1MGEA
236
EUR
B8806-1MG
BCECF AM ester 1 mg/mL in DMSO fluoreszierender pH-Indikator, Sigma-Aldrich®
BCECF AM ester
An intracellular fluorescent pH indicator.
Mixture of six isomeric compounds
Formel:
C₄₂H₄₀O₂₁ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036969 CAS-Nummer: 117464-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...