Biotin ethylenediamine hydrobromide ≥95% (durch TLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: N-(2-Aminoethyl)biotinamide hydrobromide, Neurobiotin™, (3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide hydrobromide

B9181-25MG
B9181-25MGEA 272 EUR
B9181-25MG
Biotin ethylenediamine hydrobromide ≥95% (durch TLC), Sigma-Aldrich®
Biotin ethylenediamine hydrobromide

Biotin ethylenediamine hydrobromide. ≥95% (TLC), solid ((3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide; N-(2-Aminoethyl)biotinamide hydrobromide)

Formel: C₁₂H₂₃BrN₄O₂S
Molecular Weight: 367,31 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00467807
CAS-Nummer: 216299-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c62b162a4d432d91be21a11c2418edfa401812&vid=7424933549b731c6c4d822dcff29b37226cbed73&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR