Biotin-4-fluorescein, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

B9431-5MG
B9431-5MGEA 475 EUR
B9431-5MG
Biotin-4-fluorescein, Sigma-Aldrich®
Biotin-4-fluorescein

Useful for quantitating biotin binding sites. This reagent binds to avidin more quickly than conventional fluorescein biotin.

Formel: C₃₃H₃₂N₄O₈S
Molecular Weight: 644,7 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1032732-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=080f58f8ebdb491c1ca4b7c084eb67419259b68e&vid=9c6e941cdafbe62d2f1f695abc16353457c6528d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR