7-Benzyloxygramin, Supelco®

Lieferant: Merck

B9875-100MG
B9875-100MGEA 144 EUR
B9875-100MG
7-Benzyloxygramin, Supelco®
7-Benzyloxygramin
Formel: C₁₈H₂₀N₂O
CAS-Nummer: 94067-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcfb1abe7a6e001762c8599c519b79752d39b984&vid=dcfb1abe7a6e001762c8599c519b79752d39b984&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR