N-Boc-L-tyrosin

Lieferant: BACHEM BIOCHEMICA

Synonyme: BOC-L-Tyrosine, Boc-Tyr-OH

4000198.0025 4000198.0100
BACE4000198.0025EA 0 EUR
BACE4000198.0025 BACE4000198.0100
N-Boc-L-tyrosin
N-Boc-L-tyrosin
Formel: C₁₄H₁₉NO₅
MDL: MFCD00037179
CAS-Nummer: 3978-80-1
EINECS: 223-613-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR