N(α)-Boc-L-tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-Trp-OH

4000240.0100
BACE4000240.0100EA 0 EUR
BACE4000240.0100
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304.3 g/mol
MDL: MFCD00065595
CAS-Nummer: 13139-14-5
EINECS: 236-072-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR