N-Boc-L-valin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-Val-OH, BOC-L-Valine

4000426.0100 4000426.1000 4000426.0500
BACE4000426.0100EA 0 EUR
BACE4000426.0100 BACE4000426.1000 BACE4000426.0500
N-Boc-L-valin
N-Boc-L-valin
Formel: C₁₀H₁₉NO₄
MDL: MFCD00065605
CAS-Nummer: 13734-41-3
EINECS: 237-307-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR