4-Fluor-DL-phenylalanin
Lieferant: BACHEM BIOCHEMICA
4000519.0005
BACE4000519.0005EA
0
EUR
BACE4000519.0005
4-Fluor-DL-phenylalanin
4-Fluor-DL-phenylalanin
Formel:
C₉H₁₀FNO₂ Molecular Weight: 183,18 g/mol |
MDL:
MFCD00002600 CAS-Nummer: 51-65-0 EINECS: 200-113-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...