N(α)-Boc-L-histidin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-His-OH

4000667.0005 4000667.0025 4000667.0100
BACE4000667.0005EA 0 EUR
BACE4000667.0005 BACE4000667.0025 BACE4000667.0100
N(α)-Boc-L-histidin
N(α)-Boc-L-histidin
Formel: C₁₁H₁₇N₃O₄
MDL: MFCD00065576
CAS-Nummer: 17791-52-5
EINECS: 241-768-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR