N-Acetyl-D-leucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: N-Acetyle-D-Leucine, N-Ac-D-Leu-OH, Ac-D-Leu-OH, 2-(Acetylamino)-4-methylpentanoic acid

4001881.0001
BACE4001881.0001EA 0 EUR
BACE4001881.0001
N-Acetyl-D-leucin
N-Acetyl-D-leucin
Formel: C₈H₁₅NO₃
Molecular Weight: 173,2 g/mol
MDL: MFCD00066069
CAS-Nummer: 19764-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR