N(α)-Fmoc-D-tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: Fmoc-D-Trp-OH

4003200.0005
BACE4003200.0005EA 0 EUR
BACE4003200.0005
N(α)-Fmoc-D-tryptophan
N(α)-Fmoc-D-tryptophan
Formel: C₂₆H₂₂N₂O₄
Molecular Weight: 426.46 g/mol
MDL: MFCD00062954
CAS-Nummer: 86123-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR