N(α)-Boc-D-tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-D-Trp-OH

4003847.0025
BACE4003847.0025EA 0 EUR
BACE4003847.0025
N(α)-Boc-D-tryptophan
N(α)-Boc-D-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304.3 g/mol
MDL: MFCD00037944
CAS-Nummer: 5241-64-5
EINECS: 226-042-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR