N(α)-Acetyl-L-Tryptophan
Lieferant: BACHEM BIOCHEMICA
4003876.0025
BACE4003876.0025EA
0
EUR
BACE4003876.0025
N(α)-Acetyl-L-Tryptophan
N(α)-Acetyl-L-tryptophan
Formel:
C₁₃H₁₄N₂O₃ |
MDL:
MFCD00065976 CAS-Nummer: 1218-34-4 EINECS: 214-935-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...