Ac-D-Pro-OH
Lieferant: BACHEM BIOCHEMICA
4004420.0001
BACE4004420.0001EA
0
EUR
BACE4004420.0001
Ac-D-Pro-OH
Ac-D-Pro-OH
Formel:
C₇H₁₁NO₃ |
MDL:
MFCD00080841 CAS-Nummer: 59785-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...