N-Acetyl-L-leucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: N-Ac-Leu-OH, Ac-Leu-OH

4004426.0025
BACE4004426.0025EA 0 EUR
BACE4004426.0025
N-Acetyl-L-leucin
N-Acetyl-L-leucin
Formel: C₈H₁₅NO₃
Molecular Weight: 173.2 g/mol
MDL: MFCD00065131
CAS-Nummer: 1188-21-2
EINECS: 214-706-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR