9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)

Lieferant: BACHEM BIOCHEMICA

Synonyme: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

4004776.0025
BACE4004776.0025EA 0 EUR
BACE4004776.0025
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
Formel: C₁₅H₁₁ClO₂
Molecular Weight: 258,7 g/mol
MDL: MFCD00001138
CAS-Nummer: 28920-43-6
EINECS: 249-313-6
UN: 2923
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR