DL-tert-Leucin (H-DL-Tle-OH)

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-DL-Tle-OH

4005657.0001
BACE4005657.0001EA 0 EUR
BACE4005657.0001
DL-tert-Leucin (H-DL-Tle-OH)
DL-tert-Leucin (H-DL-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131,2 g/mol
MDL: MFCD00065933
CAS-Nummer: 33105-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR