N(α)-Boc-D-histidin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-D-His-OH

4007022.0005
BACE4007022.0005EA 0 EUR
BACE4007022.0005
N(α)-Boc-D-histidin
N(α)-Boc-D-histidin
Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255.3 g/mol
MDL: MFCD00037851
CAS-Nummer: 50654-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR