L(+)-Norleucin

Lieferant: BACHEM BIOCHEMICA
Achtung

4007429.0005 4007429.0025
BACE4007429.0005EA 0 EUR
BACE4007429.0005 BACE4007429.0025
L(+)-Norleucin
L(+)-Norleucin
Formel: C₆H₁₃NO₂
Siedepunkt: 246 °C (1013 hPa)
Schmelzpunkt: 113 °C
MDL: MFCD00064423
CAS-Nummer: 327-57-1
EINECS: 206-321-4
Merck Index: 13,06738

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR