N-Boc-DL-valin

Lieferant: BACHEM BIOCHEMICA

Synonyme: BOC-DL-Valine, Boc-DL-Val-OH, 2-((tert-Butoxycarbonyl)amino)-3-methylbutanoic acid

4007725.0025
BACE4007725.0025EA 0 EUR
BACE4007725.0025
N-Boc-DL-valin
N-Boc-DL-valin
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217,26 g/mol
MDL: MFCD00069973
CAS-Nummer: 54895-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR