D-allo-Threonin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid
4008353.0250
4008353.1000
BACE4008353.0250EA
0
EUR
BACE4008353.0250
BACE4008353.1000
D-allo-Threonin
D-allo-Threonin
Formel:
C₄H₉NO₃ Molecular Weight: 119.12 g/mol |
MDL:
MFCD00004526 CAS-Nummer: 24830-94-2 EINECS: 246-488-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...