L-allo-Threonin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH

4008379.0250 4008379.1000
BACE4008379.0250EA 0 EUR
BACE4008379.0250 BACE4008379.1000
L-allo-Threonin
L-allo-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
MDL: MFCD00064268
CAS-Nummer: 28954-12-3
EINECS: 249-327-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR