4-Chlor-D-phenylalanin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Phe(4-Cl)-OH, (R)-2-Amino-3-(4-chlorophenyl)propionic acid, H-p-Chloro-D-Phe-OH

4011117.0001
BACE4011117.0001EA 0 EUR
BACE4011117.0001
4-Chlor-D-phenylalanin
4-Chlor-D-phenylalanin
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199.64 g/mol
MDL: MFCD00079675
CAS-Nummer: 14091-08-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR