3-Fluor-DL-phenylalanin

Lieferant: BACHEM BIOCHEMICA

Synonyme: 2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH

4011126.0005
BACE4011126.0005EA 151 EUR
BACE4011126.0005
3-Fluor-DL-phenylalanin
3-Fluor-DL-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,2 g/mol
MDL: MFCD00004273
CAS-Nummer: 2629-54-1
EINECS: 220-104-1
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR