N-[(Benzyloxy)carbonyl]tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: Cbz-DL-Trp-OH, Z-DL-Trp-OH

4011146.0025
BACE4011146.0025EA 0 EUR
BACE4011146.0025
N-[(Benzyloxy)carbonyl]tryptophan
N(α)-Cbz-DL-tryptophan
Formel: C₁₉H₁₈N₂O₄
Molecular Weight: 338,4 g/mol
MDL: MFCD00069707
CAS-Nummer: 13058-16-7
EINECS: 235-949-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR