Ac-DL-Pro-OH

Lieferant: BACHEM BIOCHEMICA

4013912.0005 4013912.0025 4013912.0100
BACE4013912.0005EA 0 EUR
BACE4013912.0005 BACE4013912.0025 BACE4013912.0100
Ac-DL-Pro-OH
Ac-DL-Pro-OH
Formel: C₇H₁₁NO₃
MDL: MFCD00063230
CAS-Nummer: 1074-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR