(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)

Lieferant: BACHEM BIOCHEMICA

Synonyme: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

4013924.0100
BACE4013924.0100EA 0 EUR
BACE4013924.0100
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)
Formel: C₁₂H₁₅NO₈
Lagertemperatur: Gefrierschrank
MDL: MFCD00063255
CAS-Nummer: 369-07-3
EINECS: 206-716-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR