(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate salt
Lieferant: BACHEM BIOCHEMICA
4015938.0001
BACE4015938.0001EA
0
EUR
BACE4015938.0001
(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate salt
Proteine und Peptide
The oxytocin antagonist OVTA shows enhanced affinity for rat uterine oxytocin receptors after iodination.
- Sequence: β-Mercapto-β,β-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Thr-Asn-Cys-Pro-Orn-Tyr-NH₂
- Source: synthetic
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...