(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: (3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

4016025.0005
BACE4016025.0005EA 0 EUR
BACE4016025.0005
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.20 g/mol
MDL: MFCD00144533
CAS-Nummer: 74163-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR