DL-Methioninmethylester Hydrochlorid

Lieferant: BACHEM BIOCHEMICA

Synonyme: Methyl 2-amino-4-(methylthio)butanoate hydrochloride, H-DL-Met-OMe·HCl

4016125.0025
BACE4016125.0025EA 0 EUR
BACE4016125.0025
DL-Methioninmethylester Hydrochlorid
DL-Methioninmethylester Hydrochlorid
Formel: C₆H₁₃NO₂S·HCl
Molecular Weight: 199.7 g/mol
MDL: MFCD00067540
CAS-Nummer: 16118-36-8
EINECS: 240-285-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR