DL-allo-Threonin

Lieferant: BACHEM BIOCHEMICA

4016157.0250
BACE4016157.0250EA 0 EUR
BACE4016157.0250
DL-allo-Threonin
DL-allo-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
MDL: MFCD00067248
CAS-Nummer: 144-98-9
EINECS: 205-645-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR