N-Fmoc-L-norleucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Fmoc-Nle-OH, FMOC-L-Norleucine

4018558.0001
BACE4018558.0001EA 0 EUR
BACE4018558.0001
N-Fmoc-L-norleucin
N-Fmoc-L-norleucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353.4 g/mol
MDL: MFCD00037537
CAS-Nummer: 77284-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR