Fmoc-Phenylalaninol
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Fmoc-Phenylalaninol, N-Fmoc-L-phenylalaninol
4025445.0001
BACE4025445.0001EA
0
EUR
BACE4025445.0001
Fmoc-Phenylalaninol
Fmoc-phenylalaninol
Formel:
C₂₄H₂₃NO₃ Molecular Weight: 373.452 g/mol |
MDL:
MFCD00235941 CAS-Nummer: 129397-83-7 EINECS: 252-661-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...