3-Fluor-L-phenylalanin

Lieferant: BACHEM BIOCHEMICA

4025830.0001
BACE4025830.0001EA 0 EUR
BACE4025830.0001
3-Fluor-L-phenylalanin
3-Fluor-L-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.1796 g/mol
MDL: MFCD00066450
CAS-Nummer: 19883-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR