3-Nitro-L-tyrosin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine

4026245.0025
BACE4026245.0025EA 0 EUR
BACE4026245.0025
3-Nitro-L-tyrosin
3-Nitro-L-tyrosin
Formel: C₉H₁₀N₂O₅
Molecular Weight: 226.3 g/mol
MDL: MFCD00007123
CAS-Nummer: 621-44-3
EINECS: 210-688-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR