N(α)-Acetyl-D-Tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Trp-OH, Ac-D-Trp-OH, N-Acetyl-D-tryptophan

4027317.0005
BACE4027317.0005EA 0 EUR
BACE4027317.0005
N(α)-Acetyl-D-Tryptophan
N(α)-Acetyl-D-tryptophan
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,3 g/mol
MDL: MFCD00065021
CAS-Nummer: 2280-01-5
EINECS: 218-912-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR