(S)-(+)-Thiazolidin-4-carbonsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Thz-OH, (S)-Thiazolidine-4-carboxylic acid, D-Thioproline

4027488.0005
BACE4027488.0005EA 0 EUR
BACE4027488.0005
(S)-(+)-Thiazolidin-4-carbonsäure
(S)-(+)-Thiazolidin-4-carbonsäure
Formel: C₄H₇NO₂S
Molecular Weight: 133.17 g/mol
MDL: MFCD00137425
CAS-Nummer: 45521-09-3
EINECS: 256-240-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR