N-Acetylglycin (Ac-Gly-OH)

Lieferant: BACHEM BIOCHEMICA

4030301.0500
BACE4030301.0500EA 69.6 EUR
BACE4030301.0500
N-Acetylglycin (Ac-Gly-OH)
N-Acetylglycin (Ac-Gly-OH)
Formel: C₄H₇NO₃
MDL: MFCD00004275
CAS-Nummer: 543-24-8
EINECS: 208-839-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR