D(-)-Arginin
Lieferant: BACHEM BIOCHEMICA
|
Achtung
|
Synonyme:
(R)-(-)-Arginin, H-D-Arg-OH, D(-)-α-Amino-δ-guanidinovaleriansäure, (R)-(-)-α-Amino-δ-guanidinovaleriansäure
4030317.0025
BACE4030317.0025EA
0
EUR
BACE4030317.0025
D(-)-Arginin
D(-)-Arginin
Formel:
C₆H₁₄N₄O₂ Molecular Weight: 174.2 g/mol Schmelzpunkt: 212 |
MDL:
MFCD00063116 CAS-Nummer: 157-06-2 EINECS: 205-866-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...