L(+)-Asparagin Monohydrat
Lieferant: BACHEM BIOCHEMICA
Synonyme:
(S)-(+)-Asparagin Monohydrat, (S)-(+)-2-Amino-1,4-butandisäure-4-amid Monohydrat, L-Asparagin Monohydrat, L(+)-2-Amino-1,4-butandisäure-4-amid Monohydrat, H-Asn-OH.H2O
4030374.0250
BACE4030374.0250EA
0
EUR
BACE4030374.0250
L(+)-Asparagin Monohydrat
L(+)-Asparagin Monohydrat
Formel:
C₄H₈N₂O₃·1H₂O Molecular Weight: 150.13 g/mol Schmelzpunkt: 215…217 °C Dichte: 1.567 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151038 CAS-Nummer: 5794-13-8 EINECS: 200-735-9 Merck Index: 14,00837 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...