L(-)-Cystin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (R)-(-)-Cystin, (2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure), (H-Cys-OH)2

4030444.0100
BACE4030444.0100EA 0 EUR
BACE4030444.0100
L(-)-Cystin
L(-)-Cystin
Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
Molecular Weight: 240.3 g/mol
Siedepunkt: 428
Schmelzpunkt: 261…262 °C
Dichte: 1.677 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064228
CAS-Nummer: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR