L(+)-Cystein Hydrochlorid Monohydrat

Lieferant: BACHEM BIOCHEMICA

4030461.0100
BACE4030461.0100EA 0 EUR
BACE4030461.0100
L(+)-Cystein Hydrochlorid Monohydrat
L(+)-Cystein Hydrochlorid Monohydrat
Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
Schmelzpunkt: 175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065606
CAS-Nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR