L(+)-Histidin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
(S)-(+)-2-Amino-3-(4-imidazoyl)propansäure, H, (S)-(+)-Histidin, His, (2S)-(+)-Histidin, H-His-OH, L-3-Imidazol-4-ylalanin
4030670.0250
BACE4030670.0250EA
0
EUR
BACE4030670.0250
L(+)-Histidin
L(+)-Histidin
Formel:
C₆H₉N₃O₂ Molecular Weight: 155.16 g/mol Schmelzpunkt: 272…273 °C Dichte: 1.44 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064315 CAS-Nummer: 71-00-1 EINECS: 200-745-3 Merck Index: 13,04741 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...