Glycinethylester Hydrochlorid (H-Gly-OEt.HCl)

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride

4030677.0250
BACE4030677.0250EA 0 EUR
BACE4030677.0250
Glycinethylester Hydrochlorid (H-Gly-OEt.HCl)
Glycinethylester Hydrochlorid (H-Gly-OEt.HCl)
Formel: C₄H₁₀ClNO₂
Molecular Weight: 139.58 g/mol
MDL: MFCD00012871
CAS-Nummer: 623-33-6
EINECS: 210-787-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR