L-Methionin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Met-OH, (S)-(-)-2-Amino-4-(methylmercapto)buttersäure, (S)-(-)-2-Amino-4-(methylthio)buttersäure, L(-)-Methionin, (S)-(-)-Methionin

4030850.0250
BACE4030850.0250EA 0 EUR
BACE4030850.0250
L-Methionin
L-Methionin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.21 g/mol
Siedepunkt: 278
Schmelzpunkt: 136
Dichte: 1.373 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063097
CAS-Nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR