D(-)-Norleucin

Lieferant: BACHEM BIOCHEMICA

4031116.0005 4031116.0025
BACE4031116.0005EA 0 EUR
BACE4031116.0005 BACE4031116.0025
D(-)-Norleucin
D(-)-Norleucin
Formel: C₆H₁₃NO₂
Schmelzpunkt: 301 °C
MDL: MFCD00008099
CAS-Nummer: 327-56-0
EINECS: 206-320-9
Merck Index: 12,06800

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR